missing translation for 'onlineSavingsMsg'
Learn More

2-Heptanone, ≥99% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019411
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019411 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019411 Supplier Merck Emd Millipore Supplier No. 024761ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

CAS 110-43-0
Ignition Point 739°F
Vapor Pressure 2.14 mmHg (20°C)
Vapor Density 3.94 (vs. air)
Density 0.82 g/mL (at 25°C)
Flash Point 41°C
Linear Formula CH3(CH2)4 COCH3
MDL Number MFCD00009513
Melting Point -35°C
Boiling Point 149°C to 150°C
Synonym Methyl pentyl ketone
Type Analytical Standard
Quantity 1 mL
Formula Weight 114.19
Percent Purity ≥99% (GC)
Grade Analytical Standard
Show More Show Less