missing translation for 'onlineSavingsMsg'
Learn More

2-Heptanone 98.0+%, TCI America™

Catalog No. H0037500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0037500ML 500 mL
H003725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0037500ML Supplier TCI America Supplier No. H0037500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 110-43-0
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00009513
InChI Key CATSNJVOTSVZJV-UHFFFAOYSA-N
Synonym 2-heptanone, methyl amyl ketone, butylacetone, methyl pentyl ketone, methyl n-amyl ketone, amyl methyl ketone, n-amyl methyl ketone, n-pentyl methyl ketone, heptanone, pentyl methyl ketone
PubChem CID 8051
ChEBI CHEBI:5672
IUPAC Name heptan-2-one
SMILES CCCCCC(C)=O

Specifications

Melting Point -36°C
Color Colorless
Boiling Point 151°C
Quantity 500 mL
UN Number 1110
Formula Weight 114.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Heptanone
EINECSNumber (2)-0542
RTECSNumber MJ5075000
TSCA Yes