missing translation for 'onlineSavingsMsg'
Learn More

2-Heptanone, 98%

Catalog No. AC154002500
Change view
Click to view available options
Quantity:
100 mL
250 mL
1 L
5 L
Packaging:
Glass bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC154002500 250 mL Glass bottle
AC154001000 100 mL Glass bottle
AC154000010 1 L Glass bottle
AC154000051 5 L Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC154002500 Supplier Thermo Scientific Chemicals Supplier No. 154002500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 110-43-0 | C7H14O | 114.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 110-43-0
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00009513
InChI Key CATSNJVOTSVZJV-UHFFFAOYSA-N
Synonym 2-heptanone, methyl amyl ketone, butylacetone, methyl pentyl ketone, methyl n-amyl ketone, amyl methyl ketone, n-amyl methyl ketone, n-pentyl methyl ketone, heptanone, pentyl methyl ketone
PubChem CID 8051
ChEBI CHEBI:5672
IUPAC Name heptan-2-one
SMILES CCCCCC(C)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -35.0°C
Color Colorless
Density 0.8200g/mL
Boiling Point 149.0°C to 150.0°C
Flash Point 39°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4075 to 1.4095
Linear Formula CH3(CH2)4COCH3
Quantity 250 mL
Beilstein 01, 699
Specific Gravity 0.82
Merck Index 15, 4698
Solubility Information Solubility in water: 4.3g/L (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 114.19
Percent Purity 98%
Grade Pure
Physical Form Liquid
Chemical Name or Material 2-Heptanone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
May cause drowsiness or dizziness.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi
EINECSNumber 203-767-1

RUO – Research Use Only