accessibility menu, dialog, popup

UserName

H3COOCH3

CAS RN 110-42-9

IUPAC Name: methyl decanoate
Synonyms: methyl decanoate
Molecular Weight (g/mol): 186.3
Molecular Formula: C11H22O2
InChi Key: YRHYCMZPEVDGFQ-UHFFFAOYSA-N
SMILES: CCCCCCCCCC(=O)OC
Boiling Point 
  • (3)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (1)
  • (4)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)