missing translation for 'onlineSavingsMsg'
Learn More

Methyl Decanoate 98.0+%, TCI America™

Catalog No. D0023500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0023500ML 500 mL
D002325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0023500ML Supplier TCI America Supplier No. D0023500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 110-42-9
Molecular Formula C11H22O2
Molecular Weight (g/mol) 186.30
MDL Number MFCD00009580
InChI Key YRHYCMZPEVDGFQ-UHFFFAOYSA-N
Synonym methyl caprate, decanoic acid, methyl ester, methyl caprinate, capric acid methyl ester, decanoic acid methyl ester, methyl-n-caprate, uniphat a30, metholene 2095, methyl n-decanoate, methyl n-caprate
PubChem CID 8050
IUPAC Name methyl decanoate
SMILES CCCCCCCCCC(=O)OC

Specifications

Melting Point -18°C
Color Colorless
Boiling Point 229°C
Quantity 500 mL
Formula Weight 186.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Decanoate
EINECSNumber (2)-0782
RTECSNumber HD9830000
TSCA Yes