missing translation for 'onlineSavingsMsg'
Learn More

Methyl Decanoate 99.5+%, TCI America™

Catalog No. S03085ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03085ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S03085ML Supplier TCI America Supplier No. S03085ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 110-42-9
Molecular Formula C11H22O2
Molecular Weight (g/mol) 186.30
MDL Number MFCD00009580
InChI Key YRHYCMZPEVDGFQ-UHFFFAOYSA-N
Synonym methyl caprate, decanoic acid, methyl ester, methyl caprinate, capric acid methyl ester, decanoic acid methyl ester, methyl-n-caprate, uniphat a30, metholene 2095, methyl n-decanoate, methyl n-caprate
PubChem CID 8050
IUPAC Name methyl decanoate
SMILES CCCCCCCCCC(=O)OC

Specifications

Color Colorless
Boiling Point 229°C
Quantity 5 mL
Formula Weight 186.30
Percent Purity ≥99.5% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Decanoate [Standard Material for GC]
EINECSNumber (2)-0782
RTECSNumber HD9830000
TSCA Yes