accessibility menu, dialog, popup

UserName

CAS RN 1093878-42-2

SOONBr

CAS RN 1093878-42-2

IUPAC Name: 4-(4-bromophenyl)-1λ⁶-thiomorpholine-1,1-dione
Synonyms: 4-(4-bromophenyl)-1λ⁶-thiomorpholine-1,1-dione
Molecular Weight (g/mol): 290.18
Molecular Formula: C10H12BrNO2S
InChi Key: YDNMNPHZKMEGDR-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)N1CCS(=O)(=O)CC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results