CAS RN 109306-21-0
IUPAC Name:
[5-(hydroxydiphenylmethyl)-2-methyl-2-phenyl-1,3-dioxolan-4-yl]diphenylmethanol
Synonyms:
[5-(hydroxydiphenylmethyl)-2-methyl-2-phenyl-1,3-dioxolan-4-yl]diphenylmethanol
Molecular Weight (g/mol):
528.65
Molecular Formula:
C36H32O4
InChi Key:
QVQMFUMIQACODH-UHFFFAOYNA-N
SMILES:
CC1(OC(C(O1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
(+)-4,5-Bis[hydroxy(diphenyl)methyl]-2-methyl-2-phenyl-1,3-dioxolane, TCI America™
CAS: 109306-21-0 Molecular Formula: C36H32O4 Molecular Weight (g/mol): 528.648 MDL Number: MFCD00191611 InChI Key: QVQMFUMIQACODH-UHFFFAOYSA-N Synonym: (2R,3R)-2,3-O-(1-Phenylethylidene)-1,1,4,4-tetraphenyl-1,2,3,4-butanetetrol, (2R,3R)-1,1,4,4-Tetraphenyl-2,3-O-(1-phenylethylidene)-1,2,3,4-butanetetrol, [(4R,5R)-2-Methyl-2-phenyl-1,3-dioxolane-4,5-diyl]bis(diphenylmethanol) PubChem CID: 13634622 IUPAC Name: [5-[hydroxy(diphenyl)methyl]-2-methyl-2-phenyl-1,3-dioxolan-4-yl]-diphenylmethanol SMILES: CC1(OC(C(O1)C(C2=CC=CC=C2)(C3=CC=CC=C3)O)C(C4=CC=CC=C4)(C5=CC=CC=C5)O)C6=CC=CC=C6