missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-4,5-Bis[hydroxy(diphenyl)methyl]-2-methyl-2-phenyl-1,3-dioxolane, TCI America™
Specifications
Specifications
| Quantity | 1 g |
| Formula Weight | 528.65 |
| Chemical Name or Material | (+)-4,5-Bis[hydroxy(diphenyl)methyl]-2-methyl-2-phenyl-1,3-dioxolane |
Safety and Handling
| TSCA | Yes |