accessibility menu, dialog, popup

UserName

OO

CAS RN 1090-13-7

IUPAC Name: 5,12-dihydrotetracene-5,12-dione
Synonyms: 5,12-naphthacenedione tetracenemonoquinone 5,12-naphthacenequinone naphthacene-6,11-quinone 5,12-tetracenequinone
Molecular Weight (g/mol): 258.28
Molecular Formula: C18H10O2
InChi Key: LZPBKINTWROMEA-UHFFFAOYSA-N
SMILES: O=C1C2=CC=CC=C2C(=O)C2=C1C=C1C=CC=CC1=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results