accessibility menu, dialog, popup

UserName

CH3Br

CAS RN 109-65-9

IUPAC Name: 1-bromobutane
Synonyms: 1-bromobutane
Molecular Weight (g/mol): 137.02
Molecular Formula: C4H9Br
InChi Key: MPPPKRYCTPRNTB-UHFFFAOYSA-N
SMILES: CCCCBr
Percent Purity 
  • (3)
  • (2)
  • (1)
  • (5)
  • (4)
Molecular Weight (g/mol) 
  • (9)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (11)
Boiling Point 
  • (4)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (12)
  • (1)
  • (5)
  • (2)
  • (7)
  • (3)
  • (2)
  • (1)
  • (5)
  • (4)
  • (9)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (11)
  • (4)
  • (2)
  • (3)
  • (1)
  • (5)