accessibility menu, dialog, popup

UserName

CAS RN 108963-96-8

CH3H3CCH3OONOOOCH3

CAS RN 108963-96-8

IUPAC Name: 1-tert-butyl 2-methyl 5-oxopyrrolidine-1,2-dicarboxylate
Synonyms: 1-tert-butyl 2-methyl 5-oxopyrrolidine-1,2-dicarboxylate
Molecular Weight (g/mol): 243.26
Molecular Formula: C11H17NO5
InChi Key: FNTAOUUEQHKLIU-UHFFFAOYNA-N
SMILES: COC(=O)C1CCC(=O)N1C(=O)OC(C)(C)C
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)