accessibility menu, dialog, popup

UserName

CAS RN 1086386-05-1

IUPAC Name: 3-(4-chloro-3-methylphenyl)propanoic acid
Synonyms: 3-(4-chloro-3-methylphenyl)propanoic acid
Molecular Weight (g/mol): 198.65
Molecular Formula: C10H11ClO2
InChi Key: PCLIPGMIRVJYFB-UHFFFAOYSA-N
SMILES: CC1=C(Cl)C=CC(CCC(O)=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results