missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Chloro-3-methylphenyl)propionic acid, 96%

Catalog No. AAH33072ME
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH33072ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH33072ME Supplier Thermo Scientific Chemicals Supplier No. H33072ME
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1086386-05-1 | C10H11ClO2 | 198.646 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1086386-05-1
Molecular Formula C10H11ClO2
Molecular Weight (g/mol) 198.646
MDL Number MFCD11655590
InChI Key PCLIPGMIRVJYFB-UHFFFAOYSA-N
Synonym 3-4-chloro-3-methylphenyl propanoic acid, 3-4-chloro-3-methyl-phenyl-propionic acid, 3-4-chloro-3-methylphenyl propionic acid, benzenepropanoic acid, 4-chloro-3-methyl
PubChem CID 46741400
IUPAC Name 3-(4-chloro-3-methylphenyl)propanoic acid
SMILES CC1=C(C=CC(=C1)CCC(=O)O)Cl

Specifications

Melting Point 88°C to 92°C
Quantity 500 mg
Formula Weight 198.65
Percent Purity 96%
Chemical Name or Material 3-(4-Chloro-3-methylphenyl)propionic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only