accessibility menu, dialog, popup

UserName

CAS RN 108549-23-1

IUPAC Name: [bis(benzyloxy)phosphanyl]bis(propan-2-yl)amine
Synonyms: [bis(benzyloxy)phosphanyl]diisopropylamine
Molecular Weight (g/mol): 345.42
Molecular Formula: C20H28NO2P
InChi Key: ANPWLBTUUNFQIO-UHFFFAOYSA-N
SMILES: CC(C)N(C(C)C)P(OCC1=CC=CC=C1)OCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (1)
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (3)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)