CAS RN 108-95-2
Phenol (Liquid/Certified ACS), Fisher Chemical™
CAS: 108-95-2 Molecular Formula: C6H6O Molecular Weight (g/mol): 94.11 MDL Number: MFCD00002143 InChI Key: ISWSIDIOOBJBQZ-UHFFFAOYSA-N Synonym: carbolic acid,hydroxybenzene,phenic acid,phenylic acid,oxybenzene,benzenol,phenyl hydrate,monophenol,phenyl hydroxide,phenylic alcohol PubChem CID: 996 ChEBI: CHEBI:15882 IUPAC Name: phenol SMILES: OC1=CC=CC=C1
Phenol/Chloroform/Isoamyl Alcohol (25:24:1 Mixture, pH 6.7/8.0, Liq.), Fisher BioReagents™
CAS: 108-95-2 Molecular Formula: C6H6O Molecular Weight (g/mol): 94.11 MDL Number: MFCD00002143 InChI Key: ISWSIDIOOBJBQZ-UHFFFAOYSA-N Synonym: carbolic acid,hydroxybenzene,phenic acid,phenylic acid,oxybenzene,benzenol,phenyl hydrate,monophenol,phenyl hydroxide,phenylic alcohol PubChem CID: 996 ChEBI: CHEBI:15882 IUPAC Name: phenol SMILES: OC1=CC=CC=C1
Phenol Standard, Certified, 1000ppm ±10ppm (1mL = 1mg), LabChem™
Phenol Standard, C6H6O, CAS Number-108-95-2, 7732-18-5, 500mL, 99.9, 0.1, CHEBI:15882, Colorless, 1000ppm +/-10ppm (1mL - 1mg), 1g/mL, 94.11, Certified, GHS H Statement Solution is not hazardous.
Phenol Standard, 1000 ppm C6H5OH (0.100% (w/w)), Ricca Chemical
CAS: 108-95-2 Molecular Formula: C6H6O Molecular Weight (g/mol): 94.11 MDL Number: MFCD00002143 InChI Key: ISWSIDIOOBJBQZ-UHFFFAOYSA-N PubChem CID: 996 ChEBI: CHEBI:15882 IUPAC Name: phenol SMILES: OC1=CC=CC=C1
Phenol, 99%, extra pure
CAS: 108-95-2 Molecular Formula: C6H6O Molecular Weight (g/mol): 94.11 MDL Number: MFCD00002143 InChI Key: ISWSIDIOOBJBQZ-UHFFFAOYSA-N Synonym: carbolic acid,hydroxybenzene,phenic acid,phenylic acid,oxybenzene,benzenol,phenyl hydrate,monophenol,phenyl hydroxide,phenylic alcohol PubChem CID: 996 ChEBI: CHEBI:15882 IUPAC Name: phenol SMILES: OC1=CC=CC=C1
Phenol, Saturated (pH 6.6/7.9, Liq.), Fisher BioReagents™
CAS: 108-95-2 Molecular Formula: C6H6O Molecular Weight (g/mol): 94.11 MDL Number: MFCD00002143 InChI Key: ISWSIDIOOBJBQZ-UHFFFAOYSA-N Synonym: carbolic acid,hydroxybenzene,phenic acid,phenylic acid,oxybenzene,benzenol,phenyl hydrate,monophenol,phenyl hydroxide,phenylic alcohol PubChem CID: 996 ChEBI: CHEBI:15882 IUPAC Name: phenol SMILES: OC1=CC=CC=C1
Phenol, detached crystals, 99+%
CAS: 108-95-2 Molecular Formula: C6H6O Molecular Weight (g/mol): 94.11 MDL Number: MFCD00002143 InChI Key: ISWSIDIOOBJBQZ-UHFFFAOYSA-N Synonym: carbolic acid,hydroxybenzene,phenic acid,phenylic acid,oxybenzene,benzenol,phenyl hydrate,monophenol,phenyl hydroxide,phenylic alcohol PubChem CID: 996 ChEBI: CHEBI:15882 IUPAC Name: phenol SMILES: OC1=CC=CC=C1
Phenol, ACS, 99+%, stab.
CAS: 108-95-2 Molecular Formula: C6H6O Molecular Weight (g/mol): 94.11 MDL Number: MFCD00002143 InChI Key: ISWSIDIOOBJBQZ-UHFFFAOYSA-N Synonym: carbolic acid,hydroxybenzene,phenic acid,phenylic acid,oxybenzene,benzenol,phenyl hydrate,monophenol,phenyl hydroxide,phenylic alcohol PubChem CID: 996 ChEBI: CHEBI:15882 IUPAC Name: phenol SMILES: OC1=CC=CC=C1
Phenol, 99.5%, extra pure, loose crystals, unstabilized
CAS: 108-95-2 Molecular Formula: C6H6O Molecular Weight (g/mol): 94.11 MDL Number: MFCD00002143 InChI Key: ISWSIDIOOBJBQZ-UHFFFAOYSA-N Synonym: carbolic acid,hydroxybenzene,phenic acid,phenylic acid,oxybenzene,benzenol,phenyl hydrate,monophenol,phenyl hydroxide,phenylic alcohol PubChem CID: 996 ChEBI: CHEBI:15882 IUPAC Name: phenol SMILES: OC1=CC=CC=C1
Phenol Solution, Macron Fine Chemicals™
CAS: 108-95-2 Molecular Formula: C6H6O Molecular Weight (g/mol): 94.11 MDL Number: MFCD00002143 InChI Key: ISWSIDIOOBJBQZ-UHFFFAOYSA-N Synonym: carbolic acid,hydroxybenzene,phenic acid,phenylic acid,oxybenzene,benzenol,phenyl hydrate,monophenol,phenyl hydroxide,phenylic alcohol PubChem CID: 996 ChEBI: CHEBI:15882 IUPAC Name: phenol SMILES: OC1=CC=CC=C1
Phenol, 99.5%, unstab.
CAS: 108-95-2 Molecular Formula: C6H6O Molecular Weight (g/mol): 94.11 MDL Number: MFCD00002143 InChI Key: ISWSIDIOOBJBQZ-UHFFFAOYSA-N Synonym: carbolic acid,hydroxybenzene,phenic acid,phenylic acid,oxybenzene,benzenol,phenyl hydrate,monophenol,phenyl hydroxide,phenylic alcohol PubChem CID: 996 ChEBI: CHEBI:15882 IUPAC Name: phenol SMILES: OC1=CC=CC=C1
Phenol, 99.5%, extra pure, loose crystals, unstabilized
CAS: 108-95-2 Molecular Formula: C6H6O Molecular Weight (g/mol): 94.11 MDL Number: MFCD00002143 InChI Key: ISWSIDIOOBJBQZ-UHFFFAOYSA-N Synonym: carbolic acid,hydroxybenzene,phenic acid,phenylic acid,oxybenzene,benzenol,phenyl hydrate,monophenol,phenyl hydroxide,phenylic alcohol PubChem CID: 996 ChEBI: CHEBI:15882 IUPAC Name: phenol SMILES: OC1=CC=CC=C1
Thermo Scientific Chemicals Phenol/TRIS saturated sol., for molecular biology, stabilized, DNAse, RNAse and Protease free
Phenol/TRIS , CAS # 96210-45-6, is a molecular biology solvent system that can be used for the isolation of DNA. | CAS: 108-95-2 | C6H6O
Thermo Scientific Chemicals Phenol, ultrapure, 99%, unstab.
CAS: 108-95-2 Molecular Formula: C6H6O Molecular Weight (g/mol): 94.11 MDL Number: MFCD00002143 InChI Key: ISWSIDIOOBJBQZ-UHFFFAOYSA-N Synonym: carbolic acid,hydroxybenzene,phenic acid,phenylic acid,oxybenzene,benzenol,phenyl hydrate,monophenol,phenyl hydroxide,phenylic alcohol PubChem CID: 996 ChEBI: CHEBI:15882 IUPAC Name: phenol SMILES: OC1=CC=CC=C1
Thermo Scientific Chemicals Phenol water saturated sol., for molecular biology, DNAse, RNAse, Protease free, stab., free of oxides
CAS: 108-95-2 Molecular Formula: C6H6O Molecular Weight (g/mol): 94.11 MDL Number: MFCD00002143 InChI Key: ISWSIDIOOBJBQZ-UHFFFAOYSA-N Synonym: carbolic acid,hydroxybenzene,phenic acid,phenylic acid,oxybenzene,benzenol,phenyl hydrate,monophenol,phenyl hydroxide,phenylic alcohol PubChem CID: 996 ChEBI: CHEBI:15882 IUPAC Name: phenol SMILES: OC1=CC=CC=C1