missing translation for 'onlineSavingsMsg'
Learn More

Phenol (Liquid/Certified ACS), Fisher Chemical™

Catalog No. A931I4
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A931I4 4 L Glass Bottle
A931I500 500 mL Glass Bottle
A931I1 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A931I4 Supplier Thermo Fisher Scientific Supplier No. A931I 4
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-95-2
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.11
MDL Number MFCD00002143
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
Synonym carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
PubChem CID 996
ChEBI CHEBI:15882
IUPAC Name phenol
SMILES OC1=CC=CC=C1

Specifications

Melting Point 42.8°C
Color Colorless
Density 1.0576g/cm³
pH 6
Boiling Point 182°C
Flash Point 79.4°C
Packaging Glass Bottle
Quantity 4 L
UN Number 2821
Residue after Evaporation 0.05% max.
Vapor Pressure 0.35mmHg at 25°C
Percent Purity ≥89%
Grade Certified
Physical Form Liquid
Chemical Name or Material Phenol
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Combustible liquid. Toxic by inhalation. Causes burns by all exposure routes. May cause central nervous system effects. Possible risks of irreversible effects. Danger of serious damage to health by prolonged exposure. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required.

NFPA

Health:4
Flammability:2
Instability:0
DOTInformation DOT Class 6.1, : Poison