missing translation for 'onlineSavingsMsg'
Learn More

Phenol Standard, 1000 ppm C6H5OH (0.100% (w/w)), Ricca Chemical

Catalog No. 57404
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
57404 120 mL Amber Glass Bottle
574016 500 mL Amber Glass Bottle
574032 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 57404 Supplier Ricca Chemical Company Supplier No. RI57404

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-95-2 , 7732-18-5
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.11
MDL Number MFCD00002143
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
PubChem CID 996
ChEBI CHEBI:15882
IUPAC Name phenol
SMILES OC1=CC=CC=C1

Specifications

Packaging Amber Glass Bottle
Concentration or Composition Notes 0.100% (w/w) C6H5OH
Equivalence 1 mL = 1 mg Phenol
Physical Form Liquid
Color Colorless
Melting Point Approximately 0°C
Quantity 120 mL
Chemical Name or Material Phenol Standard