CAS RN 108-93-0
IUPAC Name:
cyclohexanol
Synonyms:
cyclohexanol naxol hydrophenol cicloesanolo hydralin cykloheksanol hexalin adronol adronal Anol
Molecular Weight (g/mol):
100.16
Molecular Formula:
C6H12O
InChi Key:
HPXRVTGHNJAIIH-UHFFFAOYSA-N
SMILES:
OC1CCCCC1
1
–
2
of
2
results
1
Promotions Available
Cyclohexanol, 99%
Cyclohexanol, 99%
CAS: 108-93-0 Molecular Formula: C6H12O Molecular Weight (g/mol): 100.161 MDL Number: MFCD00003855 InChI Key: HPXRVTGHNJAIIH-UHFFFAOYSA-N Synonym: cyclohexyl alcohol,hexahydrophenol,hydrophenol,hydroxycyclohexane,hexalin,1-cyclohexanol,hydralin,adronal,naxol,adronol PubChem CID: 7966 ChEBI: CHEBI:18099 IUPAC Name: cyclohexanol SMILES: C1CCC(CC1)O
2
Promotions Available
Cyclohexanol, 98%
Cyclohexanol, 98%
CAS: 108-93-0 Molecular Formula: C6H12O Molecular Weight (g/mol): 100.16 InChI Key: HPXRVTGHNJAIIH-UHFFFAOYSA-N Synonym: cyclohexyl alcohol,hexahydrophenol,hydrophenol,hydroxycyclohexane,hexalin,1-cyclohexanol,hydralin,adronal,naxol,adronol PubChem CID: 7966 ChEBI: CHEBI:18099 IUPAC Name: cyclohexanol SMILES: C1CCC(CC1)O