accessibility menu, dialog, popup

UserName

OHHO

CAS RN 108-46-3

IUPAC Name: benzene-1,3-diol
Synonyms: resorcinol 3-hydroxycyclohexadien-1-one caswell no. 723 nako tgg m-hydroquinone durafur developer G pelagol RS resorzin developer RS pelagol grey RS
Molecular Weight (g/mol): 110.11
Molecular Formula: C6H6O2
InChi Key: GHMLBKRAJCXXBS-UHFFFAOYSA-N
SMILES: OC1=CC(O)=CC=C1
Boiling Point 
  • (2)
  • (3)
  • (7)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (2)
  • (4)
  • (4)
  • (3)
Molecular Weight (g/mol) 
  • (4)
  • (9)
Physical Form 
  • (3)
  • (2)
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (11)
  • (3)
  • (2)
  • (3)
  • (2)
  • (11)
  • (2)
  • (3)
  • (7)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (4)
  • (4)
  • (3)
  • (4)
  • (9)
  • (3)
  • (2)
  • (3)
  • (4)
  • (3)
  • (3)
  • (1)