missing translation for 'onlineSavingsMsg'
Learn More

Resorcinol, Flake, Technical, Spectrum™ Chemical

Catalog No. 18613026
Change view
Click to view available options
Quantity:
500 g
2.5 kg
25 kg
Packaging:
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613026 500 g Poly Pail
18613024 2.5 kg Poly Pail
18613025 25 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18613026 Supplier Spectrum Chemical Mfg Cor Supplier No. R1529500GM
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-46-3

Specifications

CAS Max % 1
Boiling Point 277 to 281°C
Packaging Poly Pail
Quantity 500 g
Grade Technical
Physical Form Crystalline Powder or Lumps

CAUTION: For manufacturing or laboratory use only. Not for drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.