missing translation for 'onlineSavingsMsg'
Learn More

Resorcinol 99.0+%, TCI America™

Catalog No. R0008500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0008500G 500 g
R000825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R0008500G Supplier TCI America Supplier No. R0008500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-46-3
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.112
MDL Number MFCD00002269
InChI Key GHMLBKRAJCXXBS-UHFFFAOYSA-N
Synonym 1,3-Dihydroxybenzene
PubChem CID 5054
ChEBI CHEBI:27810
IUPAC Name benzene-1,3-diol
SMILES C1=CC(=CC(=C1)O)O

Specifications

Melting Point 111°C
pH 5.2
Boiling Point 178°C
Quantity 500 g
UN Number 2876
Formula Weight 110.11
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Resorcinol
EINECSNumber (3)-0543
RTECSNumber VG9625000
TSCA Yes