accessibility menu, dialog, popup

UserName

CAS RN 108-03-2

IUPAC Name: 1-nitropropane
Synonyms: 1-nitropropane
Molecular Weight (g/mol): 89.09
Molecular Formula: C3H7NO2
InChi Key: JSZOAYXJRCEYSX-UHFFFAOYSA-N
SMILES: CCC[N+]([O-])=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)