missing translation for 'onlineSavingsMsg'
Learn More

1-Nitropropane, 98%

Catalog No. AAA1197536
Change view
Click to view available options
Quantity:
250 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1197536 500 g
AAA1197530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1197536 Supplier Thermo Scientific Chemicals Supplier No. A1197536
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 108-03-2 | C3H7NO2 | 89.094 g/mol

The solvent of 1-Nitropropane is mainly used coatings, inks and separation processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The solvent of 1-Nitropropane is mainly used coatings, inks and separation processes.

Solubility
Soluble in water. (1.4 g/L)

Notes
Store in cool place. Keep away from oxidizing agents.

Chemical Identifiers

CAS 108-03-2
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.094
MDL Number MFCD00007407
InChI Key JSZOAYXJRCEYSX-UHFFFAOYSA-N
Synonym propane, 1-nitro, nitropropane, propane, nitro, ccris 1329, dsstox_cid_980, dsstox_rid_75902, dsstox_gsid_20980, nitro-propane, n-nitropropane, 3-nitropropane
PubChem CID 7903
ChEBI CHEBI:76261
IUPAC Name 1-nitropropane
SMILES CCC[N+](=O)[O-]

Specifications

Melting Point -108°C
Density 0.998
Boiling Point 131°C to 132°C
Flash Point 34°C (93°F)
Odor Unpleasant
Refractive Index 1.402
Linear Formula CH3CH2CH2NO2
Quantity 500 g
UN Number UN2608
Beilstein 506236
Merck Index 14,6627
Solubility Information Soluble in water. (1.4g/L)
Formula Weight 89.09
Percent Purity 98%
Chemical Name or Material 1-Nitropropane
Show More Show Less
Hazard Category H226-H302+H312-H331
Hazard Statement GHS H Statement
H226-H302-H312-H332
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P311-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: NITROPROPANES
EINECSNumber 203-544-9
RTECSNumber TZ5075000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only