accessibility menu, dialog, popup

UserName

CAS RN 1077-93-6

CH3NNHONOO

CAS RN 1077-93-6

IUPAC Name: 3-(2-methyl-5-nitro-1H-imidazol-1-yl)propan-1-ol
Synonyms: 3-(2-methyl-5-nitroimidazol-1-yl)propan-1-ol
Molecular Weight (g/mol): 185.18
Molecular Formula: C7H11N3O3
InChi Key: DUOHVNSMLSPTMI-UHFFFAOYSA-N
SMILES: CC1=NC=C(N1CCCO)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)