accessibility menu, dialog, popup

UserName

CH3OHNH3C

CAS RN 1076-74-0

IUPAC Name: 5-methoxy-2-methyl-1H-indole
Synonyms: 5-methoxy-2-methyl-1H-indole
Molecular Weight (g/mol): 161.2
Molecular Formula: C10H11NO
InChi Key: VSWGLJOQFUMFOQ-UHFFFAOYSA-N
SMILES: COC1=CC=C2NC(C)=CC2=C1
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (3)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)