accessibility menu, dialog, popup

UserName

CAS RN 1073354-18-3

BOOH3CCH3H3CH3CNNCH3H3C

CAS RN 1073354-18-3

IUPAC Name: 1-(propan-2-yl)-4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazine
Synonyms: 1-isopropyl-4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazine
Molecular Weight (g/mol): 330.28
Molecular Formula: C19H31BN2O2
InChi Key: CSORKGLMGUQQOY-UHFFFAOYSA-N
SMILES: CC(C)N1CCN(CC1)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)