accessibility menu, dialog, popup

UserName

HClNH2HN

CAS RN 1073-62-7

IUPAC Name: hydrogen benzylhydrazine chloride
Synonyms: hydrogen benzylhydrazine chloride
Molecular Weight (g/mol): 158.63
Molecular Formula: C7H11ClN2
InChi Key: PRBLRLQZOKOQCQ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NNCC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results