missing translation for 'onlineSavingsMsg'
Learn More

Benzylhydrazine Monohydrochloride 98.0+%, TCI America™

Catalog No. B13965G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B13965G 5 g
B139625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B13965G Supplier TCI America Supplier No. B13965G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1073-62-7
Molecular Formula C7H11ClN2
Molecular Weight (g/mol) 158.629
MDL Number MFCD00012921
InChI Key PRBLRLQZOKOQCQ-UHFFFAOYSA-N
PubChem CID 14084
IUPAC Name benzylhydrazine;hydrochloride
SMILES C1=CC=C(C=C1)CNN.Cl

Specifications

Melting Point 113°C
Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 158.63
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Benzylhydrazine Monohydrochloride
EINECSNumber (3)-0466
RTECSNumber MU8575000
TSCA No