CAS RN 1073-26-3
IUPAC Name:
1-(1H-pyrrol-2-yl)propan-1-one
Synonyms:
1-propanone, 1-pyrrol-2-yl-
Molecular Weight (g/mol):
123.16
Molecular Formula:
C7H9NO
InChi Key:
AEJPPSRYXGEVDT-UHFFFAOYSA-N
SMILES:
CCC(=O)C1=CC=CN1
1
–
1
of
1
results
1
2-Propionylpyrrole, 99%
CAS: 1073-26-3 Molecular Formula: C7H9NO Molecular Weight (g/mol): 123.155 MDL Number: MFCD01696449 InChI Key: AEJPPSRYXGEVDT-UHFFFAOYSA-N Synonym: 2-propionylpyrrole,1-1h-pyrrol-2-yl propan-1-one,2-pyrryl ethyl ketone,ethyl 2-pyrrolyl ketone,alpha-propionylpyrrole,1-2-pyrrolyl-1-propanone,alpha-pyrryl ethyl ketone,1-propanone, 1-pyrrol-2-yl,1-1h-pyrrol-2-yl-1-propanone,unii-hgs11xf57f PubChem CID: 61260 IUPAC Name: 1-(1H-pyrrol-2-yl)propan-1-one SMILES: CCC(=O)C1=CC=CN1