accessibility menu, dialog, popup

UserName

CAS RN 107023-66-5

IUPAC Name: 2-[methyl(4-nitrophenyl)amino]acetonitrile
Synonyms: 2-[methyl(4-nitrophenyl)amino]acetonitrile
Molecular Weight (g/mol): 191.19
Molecular Formula: C9H9N3O2
InChi Key: DJOYTAUERRJRAT-UHFFFAOYSA-N
SMILES: CN(CC#N)C1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results