accessibility menu, dialog, popup

UserName

Enjoy Up to 50% Off Spotlight Deals All Year Long

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 1070-62-8

H3COOOHO

CAS RN 1070-62-8

IUPAC Name: 5-ethoxy-5-oxopentanoic acid
Synonyms: 5-ethoxy-5-oxopentanoic acid
Molecular Weight (g/mol): 160.17
Molecular Formula: C7H12O4
InChi Key: MYMNBFURSYZQBR-UHFFFAOYSA-N
SMILES: CCOC(=O)CCCC(O)=O
Boiling Point 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.