missing translation for 'onlineSavingsMsg'
Learn More

Monoethyl Glutarate 98.0+%, TCI America™

Catalog No. G027125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G027125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G027125G Supplier TCI America Supplier No. G027125G
Only null left

Chemical Identifiers

CAS 1070-62-8
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.169
MDL Number MFCD00014379
InChI Key MYMNBFURSYZQBR-UHFFFAOYSA-N
Synonym ethyl hydrogen glutarate, glutaric acid monoethyl ester, 5-ethoxy-5-oxo-pentanoic acid, ethyl glutarate,mono, acmc-20akx9, ksc493i1d, pentanedioic acid 1-ethyl ester, pentanedioic acid monoethyl ester, 5-ethoxy-5-oxidanylidene-pentanoic acid, glutaric acid monoethyl ester; monoethyl glutarate
PubChem CID 66114
IUPAC Name 5-ethoxy-5-oxopentanoic acid
SMILES CCOC(=O)CCCC(=O)O

Specifications

Color Colorless
Boiling Point 170°C
Quantity 25 g
Formula Weight 160.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Monoethyl Glutarate