accessibility menu, dialog, popup

UserName

CAS RN 107-94-8

IUPAC Name: 3-chloropropanoic acid
Synonyms: 3-chloropropionic acid
Molecular Weight (g/mol): 108.52
Molecular Formula: C3H5ClO2
InChi Key: QEYMMOKECZBKAC-UHFFFAOYSA-N
SMILES: OC(=O)CCCl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)