missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropropionic Acid 98.0+%, TCI America™

Catalog No. C0276500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0276500G 500 g
C027625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0276500G Supplier TCI America Supplier No. C0276500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 107-94-8
Molecular Formula C3H5ClO2
Molecular Weight (g/mol) 108.52
MDL Number MFCD00002764
InChI Key QEYMMOKECZBKAC-UHFFFAOYSA-N
Synonym 3-chloropropionic acid, propanoic acid, 3-chloro, propionic acid, 3-chloro, beta-chloropropionic acid, 3-chloropropanic acid, beta-monochloropropionic acid, unii-r5j180fn9z, 3-chloro-propionic acid, .beta.-chloropropionic acid, .beta.-monochloropropionic acid
PubChem CID 7899
IUPAC Name 3-chloropropanoic acid
SMILES OC(=O)CCCl

Specifications

Melting Point 40°C
Color White
Boiling Point 200°C
Quantity 500 g
UN Number 3261
Formula Weight 108.52
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloropropionic Acid
EINECSNumber (2)-1157
RTECSNumber UE8750000
TSCA Yes
Recommended Storage Refrigerator