accessibility menu, dialog, popup

UserName

CAS RN 107-91-5

IUPAC Name: 2-cyanoacetamide
Synonyms: cyanoacetamide
Molecular Weight (g/mol): 84.08
Molecular Formula: C3H4N2O
InChi Key: DGJMPUGMZIKDRO-UHFFFAOYSA-N
SMILES: NC(=O)CC#N
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (5)