missing translation for 'onlineSavingsMsg'
Learn More

Cyanoacetamide, 99%

Catalog No. AAA1248236
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1248236 500 g
AAA1248222 100 g
AAA124820E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1248236 Supplier Thermo Scientific Chemicals Supplier No. A1248236
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 107-91-5 | C3H4N2O | 84.08 g/mol

Cyanoacetamide was used in spectrofluorimetric method for the determination of some antihistaminic H1 receptor antagonist drugs such as ebastine, cetirizine dihydrochloride and fexofenadine hydrochloride. It may be used in fluorometric determination of 3,4-dihydroxyphenylalanine. It was used as post column fluorigenic derivatizing agent for the bio-analysis and pharmacokinetics of chitosan ester in rabbit serum. Cyanoacetamide is the starting reagent for vitamin B6 synthesis. It reacts with reducing carbohydrates in borate buffer to give intense fluorescence andis useful for the automated analysis of carbohydrates as borate complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyanoacetamide was used in spectrofluorimetric method for the determination of some antihistaminic H1 receptor antagonist drugs such as ebastine, cetirizine dihydrochloride and fexofenadine hydrochloride. It may be used in fluorometric determination of 3,4-dihydroxyphenylalanine. It was used as post column fluorigenic derivatizing agent for the bio-analysis and pharmacokinetics of chitosan ester in rabbit serum. Cyanoacetamide is the starting reagent for vitamin B6 synthesis. It reacts with reducing carbohydrates in borate buffer to give intense fluorescence andis useful for the automated analysis of carbohydrates as borate complexes.

Solubility
Soluble in water.

Notes
Store under cool dry place. Ensure good ventilation. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 107-91-5
Molecular Formula C3H4N2O
Molecular Weight (g/mol) 84.08
MDL Number MFCD00008024
InChI Key DGJMPUGMZIKDRO-UHFFFAOYSA-N
Synonym cyanoacetamide, acetamide, 2-cyano, cyanacetamide, malonamide nitrile, malonamonitrile, nitrilomalonamide, cyanoiminoacetic acid, 3-nitrilo-propionamide, propionamide, 3-nitrilo, usaf kf-14
PubChem CID 7898
IUPAC Name 2-cyanoacetamide
SMILES NC(=O)CC#N

Specifications

Melting Point 118°C to 122°C
Flash Point 215°C (419°F)
Quantity 500 g
Beilstein 878221
Merck Index 14,2688
Solubility Information Soluble in water.
Formula Weight 84.08
Percent Purity 99%
Chemical Name or Material Cyanoacetamide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 203-531-8
RTECSNumber AB5950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only