accessibility menu, dialog, popup

UserName

CH3OH3CCH3CH3O

CAS RN 107-70-0

IUPAC Name: 4-methoxy-4-methylpentan-2-one
Synonyms: 4-methoxy-4-methyl-2-pentanone
Molecular Weight (g/mol): 130.19
Molecular Formula: C7H14O2
InChi Key: KOKPBCHLPVDQTK-UHFFFAOYSA-N
SMILES: COC(C)(C)CC(C)=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)