missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-4-methyl-2-pentanone, 97%

Catalog No. AAB2107614
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2107614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2107614 Supplier Thermo Scientific Chemicals Supplier No. B2107614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 107-70-0 | C7H14O2 | 130.187 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 107-70-0
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
MDL Number MFCD00043932
InChI Key KOKPBCHLPVDQTK-UHFFFAOYSA-N
Synonym 4-methoxy-4-methyl-2-pentanone, pentoxone, pent-oxone solvent, 2-pentanone, 4-methoxy-4-methyl, 4-methyl-4-methoxy-2-pentanone, me 6 k, ccris 4840, acmc-1bwvk, dsstox_cid_5557, dsstox_rid_77828
PubChem CID 7884
IUPAC Name 4-methoxy-4-methylpentan-2-one
SMILES CC(=O)CC(C)(C)OC

Specifications

Density 0.91
Boiling Point 156°C
Flash Point 48°C (118°F)
Odor Mild
Refractive Index 1.418
Quantity 25 g
UN Number UN2293
Formula Weight 130.19
Percent Purity 97%
Chemical Name or Material 4-Methoxy-4-methyl-2-pentanone
Hazard Category H226-H332
Hazard Statement GHS H Statement
H226-H332
Flammable liquid and vapour.
Harmful if inhaled.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: 4-METHOXY-4-METHYLPENTAN-2-ONE
EINECSNumber 203-512-4
RTECSNumber SA7000000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only