accessibility menu, dialog, popup

UserName

CAS RN 107-12-0

IUPAC Name: propanenitrile
Synonyms: propionitrile
Molecular Weight (g/mol): 55.08
Molecular Formula: C3H5N
InChi Key: FVSKHRXBFJPNKK-UHFFFAOYSA-N
SMILES: CCC#N
Boiling Point 
  • (3)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)