missing translation for 'onlineSavingsMsg'
Learn More

Propionitrile, 99%

Catalog No. AAA13203AU
Change view
Click to view available options
Quantity:
250 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA13203AU 1000 mL
AAA13203AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA13203AU Supplier Thermo Scientific Chemicals Supplier No. A13203AU
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 107-12-0 | C3H5N | 55.08 g/mol

Propionitrile, act as solvents and are reacted further for various application including, Extraction solvent for fatty acids, oils and unsaturated hydrocarbons, Solvent for spinning and casting, removing agent of colouring matters and aromatic alcohols, Non-aqueous solvent for titrations and for inorganic salts, Recrystallization of steroids, Parent compound for organic synthesis, chemical intermediate in biochemistry, Catalyst and component of transition-metal complex catalysts, Stabilizer for chlorinated solvents, Chemical intermediate and solvent for perfumes and pharmaceuticals. It is used as a dielectric fluid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propionitrile, act as solvents and are reacted further for various application including, Extraction solvent for fatty acids, oils and unsaturated hydrocarbons, Solvent for spinning and casting, removing agent of colouring matters and aromatic alcohols, Non-aqueous solvent for titrations and for inorganic salts, Recrystallization of steroids, Parent compound for organic synthesis, chemical intermediate in biochemistry, Catalyst and component of transition-metal complex catalysts, Stabilizer for chlorinated solvents, Chemical intermediate and solvent for perfumes and pharmaceuticals. It is used as a dielectric fluid.

Solubility
It decomposes with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.Take measures to prevent the build up of electrostatic charge. Incompatible with Strong acids, Strong bases, Strong oxidizing agents, Strong reducing agents.

Chemical Identifiers

CAS 107-12-0
Molecular Formula C3H5N
Molecular Weight (g/mol) 55.08
MDL Number MFCD00001948
InChI Key FVSKHRXBFJPNKK-UHFFFAOYSA-N
Synonym propionitrile, ethyl cyanide, propiononitrile, cyanoethane, propylnitrile, propionic nitrile, ether cyanatus, hydrocyanic ether, ethylkyanid, propannitril
PubChem CID 7854
ChEBI CHEBI:26307
IUPAC Name propanenitrile
SMILES CCC#N

Specifications

Melting Point -92°C
Density 0.781
Boiling Point 97°C to 98°C
Flash Point 6°C (42°F)
Odor Pleasant
Refractive Index 1.366
Quantity 1000 mL
UN Number UN2404
Beilstein 773680
Merck Index 14,7827
Solubility Information It decomposes with water.
Formula Weight 55.08
Percent Purity 99%
Chemical Name or Material Propionitrile
Show More Show Less
Hazard Category H225-H300+H310-H319-H331
Hazard Statement GHS H Statement
H225-H300-H310-H331-H319
Highly flammable liquid and vapour.
Fatal if swallowed.
Fatal in contact with skin.
Toxic if inhaled.
Causes serious eye irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P307+P311-P310-P311-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PROPIONITRILE
EINECSNumber 203-464-4
RTECSNumber UF9625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only