accessibility menu, dialog, popup

UserName

CH3SnHOCH3OOHH3COH3C

CAS RN 1067-33-0

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 353.05
Molecular Formula: C12H26O4Sn
InChi Key: LDZDQDDROJXUBO-UHFFFAOYSA-N
SMILES: CC(O)=O.CC(O)=O.CCCC[SnH2]CCCC
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)