missing translation for 'onlineSavingsMsg'
Learn More

Dibutyltin Diacetate 95.0+%, TCI America™

Catalog No. D0302500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0302500G 500 g
D030225G 25 g
D0302100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0302500G Supplier TCI America Supplier No. D0302500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1067-33-0
Molecular Formula C12H26O4Sn
Molecular Weight (g/mol) 353.05
MDL Number MFCD00008697
InChI Key LDZDQDDROJXUBO-UHFFFAOYSA-N
SMILES CC(O)=O.CC(O)=O.CCCC[SnH2]CCCC

Specifications

Color Yellow
Boiling Point 145°C
Quantity 500 g
UN Number 2922
Formula Weight 351.03
Percent Purity ≥95.0% (W)
Physical Form Liquid
Chemical Name or Material Dibutyltin Diacetate
EINECSNumber (2)-2330
RTECSNumber WH6880000
TSCA Yes