accessibility menu, dialog, popup

UserName

CH3OHOO

CAS RN 106685-40-9

IUPAC Name: 6-[3-(adamantan-1-yl)-4-methoxyphenyl]naphthalene-2-carboxylic acid
Synonyms: adapalene allphar brand of adapalene adaferin adapaleno differine adapalene preparation alcon brand of adapalene adapalenum galderma brand of adapalene adapalene (substance)
Molecular Weight (g/mol): 412.53
Molecular Formula: C28H28O3
InChi Key: LZCDAPDGXCYOEH-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1C12CC3CC(CC(C3)C1)C2)C1=CC=C2C=C(C=CC2=C1)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)