missing translation for 'onlineSavingsMsg'
Learn More

Adapalene 98.0+%, TCI America™

Catalog No. A2549200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2549200MG 200 mg
A25491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A2549200MG Supplier TCI America Supplier No. A2549200MG
Only null left

Chemical Identifiers

CAS 106685-40-9
Molecular Formula C28H28O3
Molecular Weight (g/mol) 412.529
MDL Number MFCD03106112
InChI Key LZCDAPDGXCYOEH-UHFFFAOYSA-N
Synonym adapalene, differin, adapalenum, adapaleno, adapaleno inn-spanish, adaferin, 6-3-adamantan-1-yl-4-methoxyphenyl-2-naphthoic acid, adapalenum inn-latin, differine, 6-3-1-adamantyl-4-methoxyphenyl-2-naphthoic acid
PubChem CID 60164
ChEBI CHEBI:31174
IUPAC Name 6-[3-(1-adamantyl)-4-methoxyphenyl]naphthalene-2-carboxylic acid
SMILES COC1=C(C=C(C=C1)C2=CC3=C(C=C2)C=C(C=C3)C(=O)O)C45CC6CC(C4)CC(C6)C5

Specifications

Melting Point 324°C
Color White-Yellow
Quantity 200 mg
Formula Weight 412.53
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Adapalene
RTECSNumber QJ1987000
TSCA No
Recommended Storage Refrigerator