missing translation for 'onlineSavingsMsg'
Learn More

Adapalene Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012398
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012398 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012398 Supplier Merck Emd Millipore Supplier No. PHR1388500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Adapalene is a third-generation synthetic retinoid and a highly lipophilic compound derived from naphthoic acid. It is widely used in the treatment of acne.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 106685-40-9
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C28H28O3
MDL Number MFCD03106112
Synonym Adapalene; 6-(4-Methoxy-3-tricyclo[3.3.1.13,7 ]dec-1-ylphenyl)-2-naphthalenecarboxylic acid; 6-[3-(1-Adamantyl)-4-methoxyphenyl]-2-naphthoic acid; CD-271; Differin
Type Certified Reference Material
Quantity 500 mg
Formula Weight 412.52
Grade Certified Reference Material