CAS RN 106556-36-9
IUPAC Name:
6-(4,6-diphenyl-1,2-dihydro-1,3,5-triazin-2-ylidene)-3-methoxycyclohexa-2,4-dien-1-one
Synonyms:
6-(4,6-diphenyl-1H-1,3,5-triazin-2-ylidene)-3-methoxycyclohexa-2,4-dien-1-one
Molecular Weight (g/mol):
355.4
Molecular Formula:
C22H17N3O2
InChi Key:
SLVHGNKODJJKEZ-UHFFFAOYSA-N
SMILES:
COC1=CC(=O)C(C=C1)=C1NC(=NC(=N1)C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
2-(2-Hydroxy-4-methoxyphenyl)-4,6-diphenyl-1,3,5-triazine 98.0+%, TCI America™
CAS: 106556-36-9 Molecular Formula: C22H17N3O2 Molecular Weight (g/mol): 355.397 MDL Number: MFCD23102325 InChI Key: SLVHGNKODJJKEZ-UHFFFAOYSA-N PubChem CID: 53424132 IUPAC Name: 6-(4,6-diphenyl-1H-1,3,5-triazin-2-ylidene)-3-methoxycyclohexa-2,4-dien-1-one SMILES: COC1=CC(=O)C(=C2NC(=NC(=N2)C3=CC=CC=C3)C4=CC=CC=C4)C=C1