missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Hydroxy-4-methoxyphenyl)-4,6-diphenyl-1,3,5-triazine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 208°C |
| Color | Yellow |
| Quantity | 5 g |
| Formula Weight | 355.40 |
| Percent Purity | ≥98.0% (HPLC,N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-(2-Hydroxy-4-methoxyphenyl)-4,6-diphenyl-1,3,5-triazine |
Safety and Handling
| TSCA | Yes |