CAS RN 106312-36-1
CAS RN 106312-36-1
IUPAC Name:
4-methoxy-2-(trifluoromethyl)benzaldehyde
Synonyms:
4-methoxy-2-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol):
204.15
Molecular Formula:
C9H7F3O2
InChi Key:
BVPVUMRIGHMFNV-UHFFFAOYSA-N
SMILES:
COC1=CC(=C(C=O)C=C1)C(F)(F)F
1
–
1
of
1
results
1
4-Methoxy-2-(trifluoromethyl)benzaldehyde 97.0+%, TCI America™
CAS: 106312-36-1 Molecular Formula: C9H7F3O2 Molecular Weight (g/mol): 204.148 MDL Number: MFCD01091011 InChI Key: BVPVUMRIGHMFNV-UHFFFAOYSA-N Synonym: 4-methoxy-2-trifluoromethyl benzaldehyde,benzaldehyde, 4-methoxy-2-trifluoromethyl,4-formyl-3-trifluoromethyl anisole,pubchem1463,acmc-1c5gx,ksc504i7t,2-formyl-5-methoxybenzotrifluoride,4-methoxy-2-trifluoromethyl-benzaldehyde,benzaldehyde,4-methoxy-2-trifluoromethyl PubChem CID: 2778286 IUPAC Name: 4-methoxy-2-(trifluoromethyl)benzaldehyde SMILES: COC1=CC(=C(C=C1)C=O)C(F)(F)F