missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-2-(trifluoromethyl)benzaldehyde 97.0+%, TCI America™

Catalog No. M21621G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M21621G Supplier TCI America Supplier No. M21621G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 106312-36-1
Molecular Formula C9H7F3O2
Molecular Weight (g/mol) 204.148
MDL Number MFCD01091011
InChI Key BVPVUMRIGHMFNV-UHFFFAOYSA-N
Synonym 4-methoxy-2-trifluoromethyl benzaldehyde, benzaldehyde, 4-methoxy-2-trifluoromethyl, 4-formyl-3-trifluoromethyl anisole, pubchem1463, acmc-1c5gx, ksc504i7t, 2-formyl-5-methoxybenzotrifluoride, 4-methoxy-2-trifluoromethyl-benzaldehyde, benzaldehyde,4-methoxy-2-trifluoromethyl
PubChem CID 2778286
IUPAC Name 4-methoxy-2-(trifluoromethyl)benzaldehyde
SMILES COC1=CC(=C(C=C1)C=O)C(F)(F)F

Specifications

Melting Point 40°C
Color White
Quantity 1 g
Formula Weight 204.15
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Methoxy-2-(trifluoromethyl)benzaldehyde
TSCA No
Recommended Storage Refrigerator